Assay Details
Target Gene Details
Entrez Gene ID: | 68839 |
Gene Name: | ankyrin repeat domain 46 |
Gene Aliases: |
1110054N06Rik, AI314978, AI987733 |
Location: |
Chr.15:36477668-36503426 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankrd46 | NM_175134.4 | 5 | 2181 | NP_780343.1 |
| AB076382.1 | 5 | 2176 | BAD23946.1 | |
| AK004254.1 | 1 | 1189 | ||
| AK079270.1 | 3 | 1740 | BAC37594.1 | |
| AK088304.1 | 5 | 2142 | BAC40271.1 | |
| AK148049.1 | 5 | 2142 | BAE28312.1 | |
| BC086659.1 | 5 | 2148 | AAH86659.1 | |
| BC094595.1 | 5 | 2144 | AAH94595.1 |

Genomic Map