Assay Details
Target Gene Details
Entrez Gene ID: | 239420 |
Gene Name: | CUB and Sushi multiple domains 3 |
Gene Aliases: |
4930500N14Rik, mKIAA1894 |
Location: |
Chr.15:47580633-48792231 on Build GRCm38 |
Assay Gene Location: | Within Exon 71 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Csmd3 | NM_001081391.2 | 71 | 11709 | NP_001074860.2 |
| AK078076.1 | 11 | 2747 | ||
| AK122567.1 | 54 | 8892 | BAC65849.1 | |
| AK162216.1 | 9 | 1562 | BAE36794.1 |

Genomic Map