Assay Details
Target Gene Details
Entrez Gene ID: | 14429 |
Gene Name: | galanin receptor 3 |
Gene Aliases: |
Galnr3 |
Location: |
Chr.15:79041885-79043558 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Galr3 | NM_015738.2 | 1 | 70 | NP_056553.2 |
| BC131950.1 | 1 | 70 | ||
| BC131952.1 | 1 | 70 |
Target Gene Details
Entrez Gene ID: | 26912 |
Gene Name: | glycine C-acetyltransferase (2-amino-3-ketobutyrate-coenzyme A ligase) |
Gene Aliases: |
AI526977, Kbl |
Location: |
Chr.15:79030874-79042531 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gcat | NM_001161712.1 | NP_001155184.1 | ||
| AK076469.1 |
More Information
Set Membership: |
|

Genomic Map