Assay Details
Target Gene Details
Entrez Gene ID: | 14429 |
Gene Name: | galanin receptor 3 |
Gene Aliases: |
Galnr3 |
Location: |
Chr.15:79041885-79043558 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Galr3 | NM_015738.2 | 2 | 1093 | NP_056553.2 |
| BC131950.1 | 2 | 1093 | ||
| BC131952.1 | 2 | 1093 |
More Information
Set Membership: |
|

Genomic Map