Assay Details
Target Gene Details
Entrez Gene ID: | 11658 |
Gene Name: | activated leukocyte cell adhesion molecule |
Gene Aliases: |
AI853494, BEN, CD166, DM-GRASP, MuSC, SC1 |
Location: |
Chr.16:52248996-52453255 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Alcam | NM_009655.2 | 1 | 506 | NP_033785.1 |
AB003026.1 | 1 | 168 | BAD18062.1 | |
AK030851.1 | 1 | 507 | BAC27159.1 | |
AK053201.1 | 1 | 507 | ||
AY644766.1 | 1 | 163 | AAV28820.1 | |
BC027280.1 | 1 | 163 | AAH27280.1 | |
U95030.1 | AAC06342.1 |
