Assay Details
Target Gene Details
Entrez Gene ID: | 11658 |
Gene Name: | activated leukocyte cell adhesion molecule |
Gene Aliases: |
AI853494, BEN, CD166, DM-GRASP, MuSC, SC1 |
Location: |
Chr.16:52248996-52453255 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Alcam | NM_009655.2 | 16 | 2753 | NP_033785.1 |
AK030851.1 | 16 | 2754 | BAC27159.1 | |
AK161090.1 | 3 | 667 | BAE36190.1 | |
BC027280.1 | 16 | 2410 | AAH27280.1 |
