Assay Details
Target Gene Details
Entrez Gene ID: | 18002 |
Gene Name: | neural precursor cell expressed, developmentally down-regulated gene 8 |
Gene Aliases: |
Rub1 |
Location: |
Chr.14:55662267-55671906 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nedd8 | NM_008683.3 | 4 | 366 | NP_032709.1 |
| AK146732.1 | 4 | 367 | BAE27394.1 | |
| AK147080.1 | 4 | 351 | BAE27660.1 | |
| AK158993.1 | 3 | 233 | BAE34759.1 | |
| BC004625.1 | 4 | 351 | AAH04625.1 | |
| D10918.1 | BAA01719.1 |

Genomic Map