Assay Details
Target Gene Details
Entrez Gene ID: | 497071 |
Gene Name: | ribonuclease, RNase A family, 13 (non-active) |
Gene Aliases: |
- |
Location: |
Chr.14:51921365-51923277 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rnase13 | NM_001011687.2 | 1 | 326 | NP_001011687.1 |
BC132614.1 | 1 | 326 | ||
BC132616.1 | 1 | 326 |
