Assay Details
Target Gene Details
Entrez Gene ID: | 19752 |
Gene Name: | ribonuclease, RNase A family, 1 (pancreatic) |
Gene Aliases: |
AI574248, Rib-1, Rib1 |
Location: |
Chr.14:51145002-51146767 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnase1 | NM_011271.2 | 2 | 366 | NP_035401.2 |
| AK075775.1 | 2 | 366 | BAC35949.1 | |
| BC030036.1 | 2 | 346 | AAH30036.1 | |
| M27814.1 | 2 | 333 | AAA40060.1 |

Genomic Map