Assay Details
Target Gene Details
Entrez Gene ID: | 18163 |
Gene Name: | catenin (cadherin associated protein), delta 2 |
Gene Aliases: |
Catnd2, Nprap, neurojugin |
Location: |
Chr.15:30172593-31029344 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctnnd2 | NM_008729.2 | 23 | 4661 | NP_032755.2 |
| AK135146.1 | 1 | 518 | BAE22439.1 | |
| AK140454.1 | 13 | 2279 | BAE24395.1 | |
| AK140675.1 | 1 | 1664 | ||
| AK140960.1 | 8 | 1536 | BAE24530.1 | |
| AK150491.1 | 1 | 480 | BAE29607.1 | |
| AK152167.1 | 1 | 480 | BAE31000.1 | |
| AK164175.1 | 4 | 1134 | ||
| AK164304.1 | 6 | 1212 | BAE37727.1 | |
| BC138260.1 | ||||
| BC138261.1 | ||||
| BC145217.1 | ||||
| U90331.1 | 23 | 4664 | AAB82409.1 |
More Information
Set Membership: |
|

Genomic Map