Assay Details
Target Gene Details
Entrez Gene ID: | 66538 |
Gene Name: | ribosomal protein S19 binding protein 1 |
Gene Aliases: |
2510038A11Rik, AI604923, S19BP |
Location: |
Chr.15:80260614-80264306 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rps19bp1 | NM_175109.3 | 4 | 745 | NP_780318.1 |
AB233218.1 | 4 | 741 | BAE53365.1 | |
AK075974.1 | 4 | 726 | BAC36088.1 | |
AV119568.1 | 1 | 183 | ||
BC038691.1 | 4 | 731 | AAH38691.1 | |
BC050115.1 | 4 | 734 | AAH50115.1 |
