Assay Details
Target Gene Details
Entrez Gene ID: | 57259 |
Gene Name: | transducer of ERBB2, 2 |
Gene Aliases: |
2900090N22Rik, 4930545K18Rik, AV071822, mKIAA1663 |
Location: |
Chr.15:81848270-81858326 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tob2 | NM_020507.3 | 2 | 2798 | NP_065253.1 |
AK078476.1 | 1 | 1639 | BAC37295.1 | |
AK122537.1 | 1 | 3997 | BAC65819.1 | |
AK143542.1 | 2 | 2775 | BAE25428.1 | |
BC011163.1 | 1 | 2680 | AAH11163.1 | |
BC094031.1 | 2 | 2729 | AAH94031.1 |
