Assay Details
Target Gene Details
Entrez Gene ID: | 77945 |
Gene Name: | retinitis pigmentosa GTPase regulator interacting protein 1 |
Gene Aliases: |
0610005A07Rik, 4930401L23Rik, 4930505G06Rik, A930002K18Rik, AA415034, nmf247 |
Location: |
Chr.14:52104106-52161339 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpgrip1 | NM_001168515.1 | 25 | 4053 | NP_001161987.1 |
| NM_023879.3 | 25 | 4665 | NP_076368.1 | |
| AK029955.1 | 12 | 2665 | BAC26697.1 | |
| AY008297.1 | 25 | 4647 | AAG22857.1 | |
| BC086692.1 | 25 | 4014 |
Target Gene Details
Entrez Gene ID: | 114741 |
Gene Name: | suppressor of Ty 16 |
Gene Aliases: |
Cdc68, Fact140, Spt16, Supt16h |
Location: |
Chr.14:52160419-52197239 on Build GRCm38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Supt16 | NM_033618.3 | 26 | 4119 | NP_291096.2 |
| AF323667.1 | 26 | 4081 | AAL04452.1 | |
| AK082901.1 | 1 | 161 |
More Information
Set Membership: |
|

Genomic Map