Assay Details
Target Gene Details
Entrez Gene ID: | 20088 |
Gene Name: | ribosomal protein S24 |
Gene Aliases: |
- |
Location: |
Chr.14:24490633-24496146 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rps24 | NM_011297.2 | 6 | 725 | NP_035427.2 |
| NM_207634.1 | 5 | 705 | NP_997517.1 | |
| NM_207635.1 | 7 | 743 | NP_997518.1 | |
| AK010856.1 | 5 | 669 | BAB27225.1 |
More Information
Set Membership: |
|

Genomic Map