Assay Details
Target Gene Details
Entrez Gene ID: | 11838 |
Gene Name: | activity regulated cytoskeletal-associated protein |
Gene Aliases: |
Arc3.1, C86064, arg3.1, mArc |
Location: |
Chr.15:74669081-74672570 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Arc | NM_001276684.1 | 1 | 1673 | NP_001263613.1 |
NM_018790.3 | 1 | 1673 | NP_061260.1 | |
AF162777.1 | 1 | 1664 | AAD43586.1 | |
AK157822.1 | 1 | 1674 | BAE34212.1 | |
AK170446.1 | BAE41804.1 | |||
BC023127.1 | 1 | 1662 | AAH23127.1 |
