Assay Details
Target Gene Details
Entrez Gene ID: | 11838 |
Gene Name: | activity regulated cytoskeletal-associated protein |
Gene Aliases: |
Arc3.1, C86064, arg3.1, mArc |
Location: |
Chr.15:74669081-74672570 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arc | NM_001276684.1 | 3 | 2752 | NP_001263613.1 |
| NM_018790.3 | 3 | 2755 | NP_061260.1 | |
| AF162777.1 | 3 | 2746 | AAD43586.1 | |
| AK157822.1 | 3 | 2754 | BAE34212.1 | |
| BC023127.1 | 3 | 2741 | AAH23127.1 |

Genomic Map