Assay Details
Target Gene Details
Entrez Gene ID: | 18285 |
Gene Name: | outer dense fiber of sperm tails 1 |
Gene Aliases: |
- |
Location: |
Chr.15:38219203-38226735 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Odf1 | NM_008757.3 | 2 | 604 | NP_032783.2 |
| BC145758.1 | 2 | 604 | ||
| CJ042343.1 | ||||
| EU791469.1 | 2 | 604 | ||
| X79446.1 | 2 | 608 | CAA55965.1 |

Genomic Map