Assay Details
Target Gene Details
Entrez Gene ID: | 13196 |
Gene Name: | ArfGAP with SH3 domain, ankyrin repeat and PH domain1 |
Gene Aliases: |
AV239055, DEF-1, Ddef1, PAP, mKIAA1249, s19 |
Location: |
Chr.15:64086840-64382919 on Build GRCm38 |
Assay Gene Location: | Within Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asap1 | NM_001276461.1 | 29 | 3756 | NP_001263390.1 |
| NM_001276462.1 | 28 | 3836 | NP_001263391.1 | |
| NM_001276463.1 | 28 | 3800 | NP_001263392.1 | |
| NM_001276467.1 | 30 | 3765 | NP_001263396.1 | |
| NM_010026.3 | 31 | 3936 | NP_034156.2 | |
| AF075461.1 | 31 | 3755 | AAC98349.1 | |
| AF075462.1 | 30 | 3584 | AAC98350.1 | |
| AK122477.1 | 29 | 3504 | BAC65759.1 | |
| AK146545.1 | BAE27250.1 | |||
| AK147689.1 | 28 | 3836 | ||
| AK159048.1 | 3 | 666 | BAE34783.1 | |
| AK166056.1 | 1 | 131 | ||
| BC002201.1 | 8 | 1556 | AAH02201.1 | |
| BC048818.1 | 28 | 3766 | AAH48818.1 | |
| BC094581.1 | 31 | 3936 | AAH94581.1 | |
| U92478.1 | 10 | 1748 | AAB82338.1 |

Genomic Map