Assay Details
Target Gene Details
Entrez Gene ID: | 19296 |
Gene Name: | plasmacytoma variant translocation 1 |
Gene Aliases: |
Gt(pU21)84Imeg, Mis-1, Mlvi-1, Pvt-1 |
Location: |
Chr.15:62037987-62250975 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pvt1 | NR_003368.1 | |||
AK020628.1 | ||||
AK043790.1 | ||||
AK169445.1 | ||||
Z12002.1 |
