Assay Details
Target Gene Details
Entrez Gene ID: | 18414 |
Gene Name: | oncostatin M receptor |
Gene Aliases: |
OSMRB |
Location: |
Chr.15:6813568-6875246 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Osmr | NM_011019.3 | 18 | 3663 | NP_035149.2 |
| AF058805.1 | 18 | 3633 | AAC40122.1 | |
| AK162684.1 | 1 | 965 | BAE37021.1 |
More Information
Set Membership: |
|

Genomic Map