Assay Details
Target Gene Details
Entrez Gene ID: | 12013 |
Gene Name: | BTB and CNC homology 1 |
Gene Aliases: |
6230421P05Rik, AI323795 |
Location: |
Chr.16:87698954-87733346 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bach1 | NM_007520.2 | 5 | 3899 | NP_031546.1 |
