Assay Details
Target Gene Details
Entrez Gene ID: | 71911 |
Gene Name: | 3-hydroxybutyrate dehydrogenase, type 1 |
Gene Aliases: |
2310032J20Rik, AI327223, Bdh |
Location: |
Chr.16:31422297-31458901 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bdh1 | NM_001122683.1 | 7 | 1214 | NP_001116155.1 |
| NM_175177.4 | 7 | 1195 | NP_780386.3 | |
| AK009575.1 | 7 | 1172 | ||
| AK076718.1 | 7 | 1168 | BAC36453.1 | |
| AK137955.1 | 7 | 1155 | BAE23523.1 | |
| AK145711.1 | 7 | 1215 | BAE26605.1 | |
| AK146321.1 | 7 | 1219 | BAE27076.1 | |
| AK146962.1 | 8 | 1264 | ||
| BC027063.1 | 3 | 670 | AAH27063.1 | |
| BC043683.1 | 7 | 1198 | AAH43683.1 | |
| BC096457.1 | 7 | 1143 | AAH96457.1 | |
| EF095210.1 |
More Information
Set Membership: |
|

Genomic Map