Assay Details
Target Gene Details
Entrez Gene ID: | 224170 |
Gene Name: | DAZ interacting protein 3, zinc finger |
Gene Aliases: |
2310047C04Rik, 2A-HUB, 6430549P11Rik, A230104G20 |
Location: |
Chr.16:48924228-48994153 on Build GRCm38 |
Assay Gene Location: | Within Exon 32 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dzip3 | NM_001110017.1 | 32 | 4524 | NP_001103487.1 |
NM_027341.2 | 31 | 3906 | NP_081617.1 | |
AK032456.1 | 1 | 2402 | ||
AK122344.1 | 7 | 4592 | BAC65626.1 | |
AK160392.1 | ||||
AK163763.1 | 1 | 1794 |
