Assay Details
Target Gene Details
Entrez Gene ID: | 11911 |
Gene Name: | activating transcription factor 4 |
Gene Aliases: |
Atf-4, C/ATF, CREB2, TAXREB67 |
Location: |
Chr.15:80255184-80257545 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atf4 | NM_001287180.1 | 3 | 272 | NP_001274109.1 |
| NM_009716.3 | 2 | 555 | NP_033846.2 | |
| AA003141.1 | 3 | 271 | ||
| AK003001.1 | 2 | 556 | BAC25014.1 | |
| AK138657.1 | 2 | 240 | BAE23736.1 | |
| AK144777.1 | 2 | 241 | BAE26060.1 | |
| AK146193.1 | 2 | 243 | BAE26967.1 | |
| AK150231.1 | 2 | 242 | ||
| AK155254.1 | 2 | 241 | BAE33149.1 | |
| AK156298.1 | 2 | 235 | BAE33662.1 | |
| AK157839.1 | 2 | 210 | BAE34220.1 | |
| AK195416.1 | 2 | 165 | ||
| BC085169.1 | 2 | 446 | AAH85169.1 | |
| L13791.1 | 1 | 60 | AAA40476.1 | |
| M94087.1 | 2 | 222 | AAA53043.1 | |
| X61507.1 | 2 | 233 | CAA43723.1 |
More Information
Set Membership: |
|

Genomic Map