Assay Details
Target Gene Details
Entrez Gene ID: | 67040 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 17 |
Gene Aliases: |
2610007K22Rik, A430025E01Rik, AI047725, C80929, Gm926, p72 |
Location: |
Chr.15:79527696-79546764 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx17 | NM_001040187.1 | 13 | 3825 | NP_001035277.1 |
NM_152806.3 | 2 | 3127 | NP_690019.2 | |
NM_199080.2 | 13 | 3831 | NP_951062.1 | |
AK011338.1 | 1 | 1016 | ||
AK079294.1 | 1 | 934 | ||
AK079442.1 | 1 | 1025 | ||
AK152910.1 | 2 | 2066 | BAE31589.1 | |
AK164451.1 | 1 | 1025 | ||
BC027758.1 | 2 | 2364 | AAH27758.2 | |
BC038378.1 | 2 | 2713 | AAH38378.1 | |
BC096036.1 | 13 | 3825 | AAH96036.1 |
