Assay Details
Target Gene Details
Entrez Gene ID: | 110253 |
Gene Name: | TRIO and F-actin binding protein |
Gene Aliases: |
AI428493, EST478828, Tara, mKIAA1662 |
Location: |
Chr.15:78947724-79005869 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Triobp | NM_001039155.1 | 8 | 2405 | NP_001034244.1 |
| NM_001039156.1 | 8 | 2405 | NP_001034245.1 | |
| NM_138579.4 | 8 | 2405 | NP_613045.3 | |
| AK155197.1 | 1 | 108 | BAE33111.1 | |
| DQ228000.1 | 8 | 2405 | ABB59556.1 | |
| DQ228001.2 | 8 | 2405 | ABB59557.2 | |
| DQ228002.2 | 8 | 2405 | ABB59558.2 | |
| DQ278602.1 | 7 | 2024 | ABB77203.1 |
More Information
Set Membership: |
|

Genomic Map