Assay Details
Target Gene Details
Entrez Gene ID: | 100384868 |
Gene Name: | predicted gene, 37013 |
Gene Aliases: |
Pcdha4-g |
Location: |
Chr.18:36952648-37841873 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm37013 | NM_001174154.2 | 1 | 2086 | NP_001167625.1 |
Target Gene Details
Entrez Gene ID: | 116731 |
Gene Name: | protocadherin alpha 1 |
Gene Aliases: |
CNR |
Location: |
Chr.18:36930285-37187657 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcdha1 | NM_054072.1 | NP_473413.1 | ||
| AY013756.1 | AAK26045.1 | |||
| BC082326.1 | AAH82326.1 |
Target Gene Details
Entrez Gene ID: | 353234 |
Gene Name: | protocadherin alpha 2 |
Gene Aliases: |
- |
Location: |
Chr.18:36939205-37187661 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcdha2 | NM_198117.2 | NP_932785.1 | ||
| AY013760.1 | AAK26049.1 | |||
| BC030897.1 | ||||
| BC141390.1 | ||||
| EU854475.1 |
Target Gene Details
Entrez Gene ID: | 192163 |
Gene Name: | protocadherin alpha 3 |
Gene Aliases: |
mKIAA0345 |
Location: |
Chr.18:36946207-37187657 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcdha3 | NM_138662.1 | NP_619603.1 | ||
| AK129120.1 | BAC97930.1 | |||
| AY013761.1 | AAK26050.1 | |||
| BC141348.1 |
Target Gene Details
Entrez Gene ID: | 12936 |
Gene Name: | protocadherin alpha 4 |
Gene Aliases: |
Cnr1, Crnr1, R75250 |
Location: |
Chr.18:36952689-37187657 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcdha4 | NM_007766.2 | 1 | 2045 | NP_031792.1 |
| AK048980.1 | 1 | 2076 | ||
| AK149282.1 | 1 | 2087 | BAE28788.1 | |
| AY013762.1 | 1 | 1968 | AAK26051.1 | |
| BC060211.1 | AAH60211.1 | |||
| D86916.1 | 1 | 2045 | BAA29045.1 |
More Information
Set Membership: |
|

Genomic Map