Assay Details
Target Gene Details
Entrez Gene ID: | 433022 |
Gene Name: | phosphatidylinositol-specific phospholipase C, X domain containing 2 |
Gene Aliases: |
AW494639, EG433022 |
Location: |
Chr.16:45959261-46010413 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plcxd2 | NM_001134480.1 | 1 | 358 | NP_001127952.1 |
| CJ104359.1 | 1 | 358 |

Genomic Map