Assay Details
Target Gene Details
Entrez Gene ID: | 56698 |
Gene Name: | phosphorylated adaptor for RNA export |
Gene Aliases: |
2810055C14Rik, 4933427L19Rik, AU018701, AU018854, D18Ertd65e, Rnuxa, p55 |
Location: |
Chr.18:56562569-56587712 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phax | NM_001162989.1 | 2 | 635 | NP_001156461.1 |
| NM_019996.4 | 2 | 745 | NP_064380.3 | |
| AJ276504.1 | 2 | 576 | CAB87994.1 | |
| AK012949.1 | 2 | 615 | ||
| AK016958.1 | 2 | 636 | ||
| AK030749.1 | 2 | 603 | BAC27117.1 | |
| BC016530.1 | 2 | 592 | ||
| BC049590.1 | 2 | 591 | AAH49590.1 | |
| BC061110.1 | 2 | 605 | AAH61110.1 |
More Information
Set Membership: |
|

Genomic Map