Assay Details
Target Gene Details
Entrez Gene ID: | 225358 |
Gene Name: | family with sequence similarity 13, member B |
Gene Aliases: |
2610024E20Rik, AW060714, AW546153 |
Location: |
Chr.18:34440571-34523046 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam13b | NM_146084.1 | 21 | 3093 | NP_666196.1 |
| AK029766.1 | 21 | 4482 | ||
| BC029030.1 | 5 | 741 | AAH29030.1 | |
| BC031465.1 | 21 | 3093 | AAH31465.1 |
Target Gene Details
Entrez Gene ID: | 53871 |
Gene Name: | polycystic kidney disease 2-like 2 |
Gene Aliases: |
TRPP5 |
Location: |
Chr.18:34409152-34442789 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pkd2l2 | NM_016927.3 | 15 | 2274 | NP_058623.2 |
| AF182033.1 | 15 | 2274 | AAF65621.1 |
More Information
Set Membership: |
|

Genomic Map