Assay Details
Target Gene Details
Entrez Gene ID: | 50766 |
Gene Name: | cysteine rich transmembrane BMP regulator 1 (chordin like) |
Gene Aliases: |
AU015004 |
Location: |
Chr.17:78200185-78376592 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crim1 | NM_015800.3 | 17 | 3539 | NP_056615.1 |
| AF168680.1 | 17 | 2991 | AAF34410.1 | |
| AK034889.1 | 5 | 767 | BAC28869.1 | |
| AK053197.1 | 15 | 2483 | ||
| AK159603.1 | 4 | 418 | BAE35222.1 | |
| BC100348.1 | 17 | 3324 | AAI00349.1 |
Target Gene Details
Entrez Gene ID: | 225020 |
Gene Name: | fasciculation and elongation protein zeta 2 (zygin II) |
Gene Aliases: |
9030616F16, D17Ertd315e |
Location: |
Chr.17:78369212-78418152 on Build GRCm38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fez2 | NM_001285949.1 | NP_001272878.1 | ||
| AK165595.1 | BAE38283.1 |
More Information
Set Membership: |
|

Genomic Map