Assay Details
Target Gene Details
Entrez Gene ID: | 224902 |
Gene Name: | scaffold attachment factor B2 |
Gene Aliases: |
AA389433, AI255170, mKIAA0138 |
Location: |
Chr.17:56560971-56584672 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Safb2 | NM_001029979.2 | 21 | 3174 | NP_001025150.2 |
AK031971.1 | 3 | 569 | BAC27631.1 | |
BC028827.1 | 15 | 1888 | AAH28827.1 | |
BC050855.1 | 21 | 3108 | AAH50855.1 |
