Assay Details
Target Gene Details
Entrez Gene ID: | 100384868 |
Gene Name: | predicted gene, 37013 |
Gene Aliases: |
Pcdha4-g |
Location: |
Chr.18:36952648-37841873 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm37013 | NM_001174154.2 | NP_001167625.1 |
Target Gene Details
Entrez Gene ID: | 103611158 |
Gene Name: | predicted gene, 38666 |
Gene Aliases: |
Pcdha6-g |
Location: |
Chr.18:36967631-37841873 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm38666 | NM_001301256.1 | NP_001288185.1 |
Target Gene Details
Entrez Gene ID: | 116731 |
Gene Name: | protocadherin alpha 1 |
Gene Aliases: |
CNR |
Location: |
Chr.18:36930285-37187657 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdha1 | NM_054072.1 | NP_473413.1 | ||
AY013756.1 | AAK26045.1 | |||
BC082326.1 | AAH82326.1 |
Target Gene Details
Entrez Gene ID: | 353234 |
Gene Name: | protocadherin alpha 2 |
Gene Aliases: |
- |
Location: |
Chr.18:36939205-37187661 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdha2 | NM_198117.2 | NP_932785.1 | ||
AY013760.1 | AAK26049.1 | |||
BC030897.1 | ||||
BC141390.1 | ||||
EU854475.1 |
Target Gene Details
Entrez Gene ID: | 192163 |
Gene Name: | protocadherin alpha 3 |
Gene Aliases: |
mKIAA0345 |
Location: |
Chr.18:36946207-37187657 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdha3 | NM_138662.1 | NP_619603.1 | ||
AK129120.1 | BAC97930.1 | |||
AY013761.1 | AAK26050.1 | |||
BC141348.1 |
Target Gene Details
Entrez Gene ID: | 12936 |
Gene Name: | protocadherin alpha 4 |
Gene Aliases: |
Cnr1, Crnr1, R75250 |
Location: |
Chr.18:36952689-37187657 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdha4 | NM_007766.2 | NP_031792.1 | ||
AK149282.1 | BAE28788.1 | |||
AY013762.1 | AAK26051.1 | |||
BC060211.1 | AAH60211.1 | |||
D86916.1 | BAA29045.1 |
Target Gene Details
Entrez Gene ID: | 12941 |
Gene Name: | protocadherin alpha 5 |
Gene Aliases: |
Cnr6, Crnr6 |
Location: |
Chr.18:36960440-37187657 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdha5 | NM_009959.1 | NP_034089.1 | ||
AB008181.1 | BAA29051.1 | |||
AY013763.1 | AAK26052.1 |
Target Gene Details
Entrez Gene ID: | 12937 |
Gene Name: | protocadherin alpha 6 |
Gene Aliases: |
CNR, Cnr2, Crnr2 |
Location: |
Chr.18:36967756-37187657 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdha6 | NM_007767.3 | NP_031793.2 | ||
AY013764.1 | AAK26053.1 | |||
BC150815.1 | ||||
BC150816.1 | ||||
D86917.1 | BAA29046.1 |
Target Gene Details
Entrez Gene ID: | 12939 |
Gene Name: | protocadherin alpha 7 |
Gene Aliases: |
Cnr4, Crnr4 |
Location: |
Chr.18:36973924-37187657 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdha7 | NM_009957.1 | 1 | 1961 | NP_034087.1 |
AB008179.1 | 1 | 1854 | BAA29049.1 | |
AY013765.1 | 1 | 1961 | AAK26054.1 | |
BC080863.1 | 1 | 2083 | AAH80863.1 |
Target Gene Details
Entrez Gene ID: | 103611159 |
Gene Name: | protocadherin alpha 7-gamma |
Gene Aliases: |
- |
Location: |
Chr.18:36973802-37841873 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdha7-g | NM_001301259.1 | 1 | 2083 | NP_001288188.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
