Assay Details
Target Gene Details
Entrez Gene ID: | 100384868 |
Gene Name: | predicted gene, 37013 |
Gene Aliases: |
Pcdha4-g |
Location: |
Chr.18:36952648-37841873 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm37013 | NM_001174154.2 | NP_001167625.1 |
Target Gene Details
Entrez Gene ID: | 103611158 |
Gene Name: | predicted gene, 38666 |
Gene Aliases: |
Pcdha6-g |
Location: |
Chr.18:36967631-37841873 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm38666 | NM_001301256.1 | NP_001288185.1 |
Target Gene Details
Entrez Gene ID: | 116731 |
Gene Name: | protocadherin alpha 1 |
Gene Aliases: |
CNR |
Location: |
Chr.18:36930285-37187657 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcdha1 | NM_054072.1 | 4 | 4341 | NP_473413.1 |
| AY013756.1 | 4 | 4341 | AAK26045.1 | |
| BC082326.1 | 5 | 4014 | AAH82326.1 |
Target Gene Details
Entrez Gene ID: | 12943 |
Gene Name: | protocadherin alpha 10 |
Gene Aliases: |
Cnr3, Cnr8, Crnr3, Crnr8, Pcdha14 |
Location: |
Chr.18:37005320-37187657 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcdha10 | NM_009961.1 | 4 | 4335 | NP_034091.1 |
| AY013757.1 | 4 | 4335 | AAK26046.1 |
Target Gene Details
Entrez Gene ID: | 12942 |
Gene Name: | protocadherin alpha 11 |
Gene Aliases: |
A830022B16Rik, Cnr7, Crnr7 |
Location: |
Chr.18:37010858-37187657 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcdha11 | NM_009960.1 | 4 | 4347 | NP_034090.1 |
| AK043822.1 | 2 | 1847 | BAC31663.1 | |
| AY013758.1 | 4 | 4347 | AAK26047.1 | |
| BC141349.1 | 5 | 3945 |
Target Gene Details
Entrez Gene ID: | 192164 |
Gene Name: | protocadherin alpha 12 |
Gene Aliases: |
Cnr5, Crnr5, Pcdha13 |
Location: |
Chr.18:37020230-37187657 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcdha12 | NM_138663.2 | 4 | 4347 | NP_619604.1 |
| AY013759.1 | 4 | 4347 | AAK26048.1 |
Target Gene Details
Entrez Gene ID: | 353234 |
Gene Name: | protocadherin alpha 2 |
Gene Aliases: |
- |
Location: |
Chr.18:36939205-37187661 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcdha2 | NM_198117.2 | 4 | 4454 | NP_932785.1 |
| AY013760.1 | 4 | 4341 | AAK26049.1 | |
| BC141390.1 | 5 | 4007 |
Target Gene Details
Entrez Gene ID: | 192163 |
Gene Name: | protocadherin alpha 3 |
Gene Aliases: |
mKIAA0345 |
Location: |
Chr.18:36946207-37187657 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcdha3 | NM_138662.1 | 4 | 4323 | NP_619603.1 |
| AK129120.1 | 4 | 4425 | BAC97930.1 | |
| AY013761.1 | 4 | 4323 | AAK26050.1 | |
| BC141348.1 | 4 | 4360 |
Target Gene Details
Entrez Gene ID: | 12936 |
Gene Name: | protocadherin alpha 4 |
Gene Aliases: |
Cnr1, Crnr1, R75250 |
Location: |
Chr.18:36952689-37187657 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcdha4 | NM_007766.2 | 4 | 4415 | NP_031792.1 |
| AY013762.1 | 4 | 4338 | AAK26051.1 | |
| BC060211.1 | 5 | 3218 | AAH60211.1 | |
| D86916.1 | 4 | 4415 | BAA29045.1 |
Target Gene Details
Entrez Gene ID: | 12941 |
Gene Name: | protocadherin alpha 5 |
Gene Aliases: |
Cnr6, Crnr6 |
Location: |
Chr.18:36960440-37187657 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcdha5 | NM_009959.1 | 4 | 4299 | NP_034089.1 |
| AY013763.1 | 4 | 4299 | AAK26052.1 |
Target Gene Details
Entrez Gene ID: | 12937 |
Gene Name: | protocadherin alpha 6 |
Gene Aliases: |
CNR, Cnr2, Crnr2 |
Location: |
Chr.18:36967756-37187657 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcdha6 | NM_007767.3 | 4 | 4335 | NP_031793.2 |
| AY013764.1 | 4 | 4335 | AAK26053.1 | |
| BC150815.1 | 5 | 4015 | ||
| BC150816.1 | 4 | 4443 | ||
| D86917.1 | 4 | 4461 | BAA29046.1 |
Target Gene Details
Entrez Gene ID: | 12939 |
Gene Name: | protocadherin alpha 7 |
Gene Aliases: |
Cnr4, Crnr4 |
Location: |
Chr.18:36973924-37187657 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcdha7 | NM_009957.1 | 4 | 4308 | NP_034087.1 |
| AY013765.1 | 4 | 4308 | AAK26054.1 |
Target Gene Details
Entrez Gene ID: | 103611159 |
Gene Name: | protocadherin alpha 7-gamma |
Gene Aliases: |
- |
Location: |
Chr.18:36973802-37841873 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcdha7-g | NM_001301259.1 | NP_001288188.1 |
Target Gene Details
Entrez Gene ID: | 353235 |
Gene Name: | protocadherin alpha 8 |
Gene Aliases: |
- |
Location: |
Chr.18:36992467-37187657 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcdha8 | NM_201243.1 | 4 | 4335 | NP_957695.1 |
| AY013766.1 | 4 | 4335 | AAK26055.1 | |
| BC139424.1 | 4 | 4431 | ||
| BC139426.1 | 4 | 4431 |
Target Gene Details
Entrez Gene ID: | 192161 |
Gene Name: | protocadherin alpha 9 |
Gene Aliases: |
- |
Location: |
Chr.18:36997880-37187657 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcdha9 | NM_138661.1 | 4 | 4434 | NP_619602.1 |
| AY013767.1 | 4 | 4434 | AAK26056.1 |
Target Gene Details
Entrez Gene ID: | 353236 |
Gene Name: | protocadherin alpha subfamily C, 1 |
Gene Aliases: |
CNRc1 |
Location: |
Chr.18:37090136-37187657 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcdhac1 | NM_001003671.1 | 4 | 4389 | NP_001003671.1 |
| AY013768.1 | 4 | 4389 | AAK26057.1 |
Target Gene Details
Entrez Gene ID: | 353237 |
Gene Name: | protocadherin alpha subfamily C, 2 |
Gene Aliases: |
CNRc2 |
Location: |
Chr.18:37143969-37187657 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcdhac2 | NM_001003672.1 | 4 | 4515 | NP_001003672.1 |
| AY013769.1 | 4 | 4515 | AAK26058.1 |
More Information
Set Membership: |
|

Genomic Map