Assay Details
Target Gene Details
Entrez Gene ID: | 240442 |
Gene Name: | ADNP homeobox 2 |
Gene Aliases: |
8430420L05Rik, BC024969, Kiaa0863, Zfp508, mKIAA0863 |
Location: |
Chr.18:80126315-80152063 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adnp2 | NM_175028.1 | 4 | 3543 | NP_778193.1 |
AB093268.1 | 4 | 3543 | BAC41452.1 | |
AK088326.1 | 1 | 117 | ||
AK160320.1 | BAE35737.1 | |||
BC024969.1 | 1 | 780 | AAH24969.1 | |
BC044898.1 | 4 | 3448 | AAH44898.1 | |
BC044904.1 | 1 | 894 | AAH44904.1 | |
BC064672.1 | 2 | 3316 | AAH64672.1 |
