Assay Details
Target Gene Details
Entrez Gene ID: | 13510 |
Gene Name: | desmoglein 1 alpha |
Gene Aliases: |
BB010119, Dsg1 |
Location: |
Chr.18:20310873-20343353 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dsg1a | NM_010079.2 | 15 | 5068 | NP_034209.2 |
| AK029294.1 | 1 | 2726 | BAC26378.1 | |
| AK037234.1 | 1 | 1941 |
More Information
Set Membership: |
|

Genomic Map