Assay Details
Target Gene Details
Entrez Gene ID: | 100384868 |
Gene Name: | predicted gene, 37013 |
Gene Aliases: |
Pcdha4-g |
Location: |
Chr.18:36952648-37841873 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm37013 | NM_001174154.2 | NP_001167625.1 |
Target Gene Details
Entrez Gene ID: | 103611158 |
Gene Name: | predicted gene, 38666 |
Gene Aliases: |
Pcdha6-g |
Location: |
Chr.18:36967631-37841873 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm38666 | NM_001301256.1 | NP_001288185.1 |
Target Gene Details
Entrez Gene ID: | 103611159 |
Gene Name: | protocadherin alpha 7-gamma |
Gene Aliases: |
- |
Location: |
Chr.18:36973802-37841873 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcdha7-g | NM_001301259.1 | NP_001288188.1 |
Target Gene Details
Entrez Gene ID: | 93892 |
Gene Name: | protocadherin beta 21 |
Gene Aliases: |
Pcdhb18, PcdhbU |
Location: |
Chr.18:37513652-37516363 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcdhb21 | NM_053146.2 | 1 | 906 | NP_444376.1 |
| AF326314.1 | 1 | 738 | AAK53239.1 | |
| AY013783.1 | 1 | 738 | AAK26072.1 | |
| BC138121.1 | 1 | 825 | ||
| BC138122.1 | 1 | 825 |
More Information
Set Membership: |
|

Genomic Map