Assay Details
Target Gene Details
Entrez Gene ID: | 70757 |
Gene Name: | 3-hydroxyacyl-CoA dehydratase 2 |
Gene Aliases: |
6330408J20Rik, AI255777, AI481689, Hcad2, Ptplb |
Location: |
Chr.16:35022421-35109190 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hacd2 | NM_023587.2 | 7 | 3121 | NP_076076.2 |
AK148143.1 | 7 | 3107 | BAE28371.1 |
