Assay Details
Target Gene Details
Entrez Gene ID: | 100043332 |
Gene Name: | ankyrin repeat domain 66 |
Gene Aliases: |
AU041848, D730048J04Rik, Gm4368 |
Location: |
Chr.17:43534173-43570729 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankrd66 |
Target Gene Details
Entrez Gene ID: | 27226 |
Gene Name: | phospholipase A2, group VII (platelet-activating factor acetylhydrolase, plasma) |
Gene Aliases: |
R75400 |
Location: |
Chr.17:43568082-43612202 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pla2g7 | NM_013737.5 | 1 | 278 | NP_038765.2 |
| AK005158.1 | BAB23849.1 | |||
| AK005210.1 | 1 | 72 | ||
| AK051454.1 | 1 | 279 | BAC34647.1 | |
| AK155678.1 | 1 | 60 | BAE33386.1 | |
| BC010726.1 | 1 | 34 | AAH10726.1 | |
| U34277.1 | AAC52274.1 |
More Information
Set Membership: |
|

Genomic Map