Assay Details
Target Gene Details
Entrez Gene ID: | 27226 |
Gene Name: | phospholipase A2, group VII (platelet-activating factor acetylhydrolase, plasma) |
Gene Aliases: |
R75400 |
Location: |
Chr.17:43568082-43612202 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pla2g7 | NM_013737.5 | 12 | 1616 | NP_038765.2 |
| AK005210.1 | 12 | 1410 | ||
| AK051454.1 | 12 | 1617 | BAC34647.1 | |
| BC010726.1 | 12 | 1372 | AAH10726.1 | |
| BF020830.1 | 1 | 308 | ||
| U34277.1 | 12 | 1362 | AAC52274.1 |
More Information
Set Membership: |
|

Genomic Map