Assay Details
Target Gene Details
Entrez Gene ID: | 20383 |
Gene Name: | serine/arginine-rich splicing factor 3 |
Gene Aliases: |
AL024116, Sfrs3, X16 |
Location: |
Chr.17:29032660-29043372 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srsf3 | NM_013663.5 | 6 | 2242 | NP_038691.1 |
| NR_036613.1 | 7 | 2698 | ||
| AK078891.1 | 6 | 2230 | BAC37445.1 |
More Information
Set Membership: |
|

Genomic Map