Assay Details
Target Gene Details
Entrez Gene ID: | 225256 |
Gene Name: | desmoglein 1 beta |
Gene Aliases: |
Dsg5 |
Location: |
Chr.18:20376835-20412180 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dsg1b | NM_181682.1 | 11 | 1791 | NP_859010.1 |
| AK036986.1 | 11 | 1899 | ||
| AY192158.1 | 11 | 1791 | AAP31152.1 | |
| BC137927.1 | 11 | 1782 |
More Information
Set Membership: |
|

Genomic Map