Assay Details
Target Gene Details
Entrez Gene ID: | 57263 |
Gene Name: | resistin like beta |
Gene Aliases: |
9030012B21Rik, Fizz2, RELMbeta, Relmb, Xcp3 |
Location: |
Chr.16:48816856-48818892 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Retnlb | NM_023881.4 | 4 | 595 | NP_076370.3 |
AF290871.1 | 4 | 597 | AAK83103.1 | |
AF323083.1 | 4 | 482 | AAG59826.1 | |
AK020240.1 | 4 | 483 | BAB32036.1 | |
BC022650.1 | 4 | 465 | AAH22650.1 | |
DQ157777.1 | AAZ82036.1 |
