Assay Details
Target Gene Details
Entrez Gene ID: | 78376 |
Gene Name: | suppressor APC domain containing 1 |
Gene Aliases: |
2810436B06Rik, G7d, Ng23 |
Location: |
Chr.17:35025959-35028571 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sapcd1 | NM_023893.4 | 4 | 709 | NP_076382.3 |
| NR_028258.1 | ||||
| AK019275.1 | 4 | 610 | BAB31642.1 | |
| BF465693.1 | ||||
| CF739520.1 |
Target Gene Details
Entrez Gene ID: | 27762 |
Gene Name: | von Willebrand factor A domain containing 7 |
Gene Aliases: |
C6orf27, D17H6S56E-3, G7c, NG37 |
Location: |
Chr.17:35016357-35026741 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vwa7 | NM_138582.1 | 17 | 3775 | NP_613048.1 |
| AF134319.1 | 17 | 3775 | AAF61401.1 | |
| AK037977.1 | 1 | 250 | ||
| BC137792.1 | 17 | 3716 | ||
| BC144911.1 | 17 | 3653 |

Genomic Map