Assay Details
Target Gene Details
Entrez Gene ID: | 94185 |
Gene Name: | tumor necrosis factor receptor superfamily, member 21 |
Gene Aliases: |
AA959878, DR6, R74815, TR7 |
Location: |
Chr.17:43016555-43089188 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfrsf21 | NM_178589.3 | 3 | 1351 | NP_848704.1 |
| AF322069.1 | 3 | 909 | AAG38115.1 | |
| AK033529.1 | 3 | 1327 | BAC28342.1 | |
| AK043823.1 | 3 | 1353 | BAC31664.1 | |
| AK134704.1 | 3 | 1322 | BAE22249.1 | |
| AY043489.1 | 3 | 909 | AAK74193.1 | |
| BC016420.1 | 3 | 1300 | AAH16420.1 |
More Information
Set Membership: |
|

Genomic Map