Assay Details
Target Gene Details
Entrez Gene ID: | 11789 |
Gene Name: | adenomatosis polyposis coli |
Gene Aliases: |
AI047805, AU020952, AW124434, CC1, Min, mAPC |
Location: |
Chr.18:34220307-34322190 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apc | NM_007462.3 | 1 | 136 | NP_031488.2 |
| AK042021.1 | 1 | 137 | BAC31134.1 | |
| AK045053.1 | 1 | 107 | BAC32198.1 | |
| AK046280.1 | 1 | 127 | ||
| AK080907.1 | 1 | 136 | BAC38073.1 |
Target Gene Details
Entrez Gene ID: | 633395 |
Gene Name: | ribosomal protein L29 pseudogene |
Gene Aliases: |
ENSMUSG00000073607 |
Location: |
Chr.18:34207775-34221772 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm10548 | NR_040534.1 | |||
| AK132217.1 | 1 | 951 | BAE21038.1 | |
| AK160850.1 |
More Information
Set Membership: |
|

Genomic Map