Assay Details
Target Gene Details
Entrez Gene ID: | 224727 |
Gene Name: | BCL2-associated athanogene 6 |
Gene Aliases: |
2410045D21Rik, AA408914, BAG-6, Bat3, D17H6S52E, G3, Scythe |
Location: |
Chr.17:35134600-35147322 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bag6 | NM_001252468.1 | 13 | 2210 | NP_001239397.1 |
| NM_001252469.1 | 14 | 2121 | NP_001239398.1 | |
| NM_057171.2 | 14 | 2264 | NP_476512.1 | |
| AK148796.1 | 13 | 2130 | BAE28666.1 | |
| AK164266.1 | 14 | 2092 | ||
| AK170134.1 | 15 | 2189 | BAE41587.1 | |
| BC026647.1 | 6 | 1005 | AAH26647.1 |
More Information
Set Membership: |
|

Genomic Map