Assay Details
Target Gene Details
Entrez Gene ID: | 224823 |
Gene Name: | ribosomal RNA processing 36 homolog (S. cerevisiae) |
Gene Aliases: |
- |
Location: |
Chr.17:46667442-46674451 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rrp36 | NM_144857.1 | 7 | 996 | NP_659106.1 |
| AK148237.1 | 7 | 1093 | BAE28429.1 | |
| BC011248.1 | 7 | 996 | AAH11248.1 |

Genomic Map