Assay Details
Target Gene Details
Entrez Gene ID: | 240025 |
Gene Name: | dapper homolog 2, antagonist of beta-catenin (xenopus) |
Gene Aliases: |
2900084M21Rik, A630024E20, Dpr2, Frd2, dapper2 |
Location: |
Chr.17:14195230-14204338 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dact2 | NM_172826.3 | 1 | 55 | NP_766414.3 |
| AK041604.1 | BAC31003.1 | |||
| AK082856.1 | ||||
| AK131775.1 | ||||
| AK141226.1 | ||||
| AY297430.2 | 1 | 318 | AAP57185.1 | |
| BC058740.1 | AAH58740.1 |

Genomic Map