Assay Details
Target Gene Details
Entrez Gene ID: | 13163 |
Gene Name: | Fas death domain-associated protein |
Gene Aliases: |
- |
Location: |
Chr.17:33909381-33915590 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Daxx | NM_001199733.1 | 3 | 1051 | NP_001186662.1 |
| NM_007829.4 | 3 | 951 | NP_031855.3 | |
| AF006040.1 | 2 | 805 | AAC53284.1 | |
| AK090356.1 | 3 | 882 | ||
| AK133863.1 | 3 | 859 | BAE21895.1 | |
| AK144332.1 | 3 | 882 | ||
| AK145906.1 | 3 | 882 | BAE26741.1 | |
| AK146743.1 | 3 | 878 | BAE27403.1 | |
| AK151279.1 | 3 | 871 | ||
| AK153149.1 | 3 | 839 | BAE31759.1 | |
| AK156425.1 | 3 | 1052 | BAE33708.1 | |
| AK172616.1 | 3 | 1052 | ||
| AY734459.1 | 1 | 518 | AAU87500.1 | |
| AY734462.1 | 1 | 60 | AAU50681.1 | |
| AY734465.1 | 1 | 429 | AAU45095.1 | |
| AY758273.1 | 1 | 433 | AAU84992.1 | |
| BC128373.1 | 3 | 864 | ||
| KJ650207.1 | 2 | 781 |

Genomic Map